1-Amino-9,10-anthraquinone

Supplier: TCI America

Synonyms: 1-Aminoanthracene-9,10-dione1-Aminoanthraquinone

A0590-500G A0590-25G
TCA0590-025GEA 44.66 USD
TCA0590-025G TCA0590-500G
1-Amino-9,10-anthraquinone
1-Amino-9,10-anthraquinone

CAS Number: 82-45-1
MDL Number: MFCD00001213
Molecular Formula: C14H9NO2
Molecular Weight: 223.23
Form: Crystal
Color: Yellow Red
Melting point (°C): 257

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 257 °C
MDL Number: MFCD00001213
CAS Number: 82-45-1
EINECS: 201-423-5
Merck Index: 13,00416
https://us.vwr.com/stibo/web/std.lang.all/80/15/28928015.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR