4-Arsanilic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: p-Arsanilic acid

A0530-25G A0530-500G
TCA0530-500GEA 236.42 USD
TCA0530-500G TCA0530-25G
4-Arsanilic acid ≥98.0% (by HPLC, titration analysis)
4-Arsanilic acid

CAS Number: 98-50-0
MDL Number: MFCD00007819
Molecular Formula: C6H8AsNO3
Molecular Weight: 217.06
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 4-Aminobenzenearsonic Acid, p-Arsanilic Acid

Formula: C₆H₈AsNO₃
MW: 217.06 g/mol
MDL Number: MFCD00007819
CAS Number: 98-50-0
EINECS: 202-674-3
UN: 3465
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/79/60/28927960.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR