Methyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 4-aminobenzoate

A0510-25G A0510-250G
TCA0510-025GEA 59.85 USD
TCA0510-025G TCA0510-250G
Methyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
Methyl 4-aminobenzoate

CAS Number: 619-45-4
MDL Number: MFCD00007891
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: 4-Aminobenzoic Acid Methyl Ester

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 114 °C
Density: 1.216 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007891
CAS Number: 619-45-4
EINECS: 210-598-7
https://us.vwr.com/stibo/web/std.lang.all/79/42/28927942.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR