o-Anisidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2-Aminoanisole
, 2-Anisidine
, 2-Methoxyaniline
A0486-25ML
A0486-500ML
TCA0486-25MLEA
27.91
USD
TCA0486-25ML
TCA0486-500ML
o-Anisidine ≥98.0% (by GC, titration analysis)
o-Anisidine
CAS Number: 90-04-0
MDL Number: MFCD00007688
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 225
Flash Point (°C): 111
Freezing point (°C): 6
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: 2-Methoxyaniline
Formula:
C₇H₉NO MW: 123.15 g/mol Boiling Pt: 225 °C Melting Pt: 5 °C Density: 1.09 (20 °C) Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007688 CAS Number: 90-04-0 EINECS: 201-963-1 UN: 2431 ADR: 6.1,III Merck Index: 13,00670 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...