Pentylbenzene ≥98.0%

Supplier: TCI America

Synonyms: 1-PhenylpentaneAmylbenzene

A0449-500ML A0449-25ML
TCA0449-500MLEA 502 USD
TCA0449-500ML TCA0449-025ML
Pentylbenzene ≥98.0%
Pentylbenzene

CAS Number: 538-68-1
MDL Number: MFCD00009502
Molecular Formula: C11H16
Molecular Weight: 148.25
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Melting point (°C): -75
Flash Point (°C): 66
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: 1-Phenylpentane

Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 205 °C
Melting Pt: –75 °C
Density: 0.86 (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00009502
CAS Number: 538-68-1
EINECS: 208-701-5
UN: 3082
ADR: 9,III
Merck Index: 13,00604
https://us.vwr.com/stibo/web/std.lang.all/78/87/28927887.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR