3-Amino-1H-1,2,4-triazole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Amitrole
CAS Number: 61-82-5
MDL Number: MFCD00005230
Molecular Formula: C2H4N4
Molecular Weight: 84.08
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 155
Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction), Bioscience, ROS/RNS-related Enzyme Inhibitors, Analytical Science, Estradiol, etc. (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Amitrole
Formula:
C₂H₄N₄ Boiling Pt: 347 °C (1013 hPa) Melting Pt: 155 °C Density: 1.138 g/cm³ (20 °C) |
MDL Number:
MFCD00053362 CAS Number: 61-82-5 EINECS: 200-521-5 Merck Index: 13,00489 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...