(4-Aminophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
4-Aminophenylacetic acid
A0393-25G
TCA0393-025GEA
100.86
USD
TCA0393-025G
(4-Aminophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(4-Aminophenyl)acetic acid
CAS Number: 1197-55-3
MDL Number: MFCD00007916
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 192
Formula:
(C₆H₄(NH₂))CH₂COOH MW: 151.16 g/mol Boiling Pt: 322 °C (1013 hPa) Melting Pt: 192 °C |
MDL Number:
MFCD00007916 CAS Number: 1197-55-3 EINECS: 214-828-7 Merck Index: 13,00462 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...