2-Aminoisobutyric acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-Amino-2-methylpropanoic acid
A0323-500G
A0323-25G
A0323-100G
TCA0323-025GEA
37.4
USD
TCA0323-025G
TCA0323-500G
TCA0323-100G
2-Aminoisobutyric acid ≥98.0% (by titrimetric analysis)
2-Aminoisobutyric acid
CAS Number: 62-57-7
MDL Number: MFCD00008049
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 335
Product Categories: Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Methylalanine
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 204 °C (1013 hPa) Melting Pt: 335 °C Density: 1.278 g/cm³ (20 °C) |
MDL Number:
MFCD00008049 CAS Number: 62-57-7 EINECS: 200-544-0 Merck Index: 13,00444 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...