(±)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
H-DL-Abu-OH
, DL-2-Aminobutyric acid
, DL-α-Amino-N-Butyric Acid
A0280-25G
TCA0280-025GEA
62.93
USD
TCA0280-025G
(±)-2-Aminobutyric acid ≥99.0% (by titrimetric analysis)
(±)-2-Aminobutyric acid
CAS Number: 2835-81-6
MDL Number: MFCD00008093
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 304
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Melting Pt: 304 °C |
MDL Number:
MFCD00008093 CAS Number: 2835-81-6 EINECS: 220-616-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...