2-Aminothiophenol ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Aminophenyl mercaptan 2-Mercaptoaniline 2-Aminobenzenethiol

A0267-25ML A0267-500ML A0267-100ML
TCA0267-100MLEA 58.37 USD
TCA0267-100ML TCA0267-25ML TCA0267-500ML
2-Aminothiophenol ≥97.0% (by titrimetric analysis)
2-Aminothiophenol

CAS Number: 137-07-5
MDL Number: MFCD00007702
Molecular Formula: C6H7NS
Molecular Weight: 125.19
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Boiling point (°C): 234
Flash Point (°C): 122
Freezing point (°C): 18
Specific Gravity (20/20): 1.17


Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes, Analytical Science, Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 2-Aminothiophenol, 2-Mercaptoaniline

Formula: C₆H₇NS
MW: 125.19 g/mol
Boiling Pt: 234 °C
Melting Pt: 26 °C
Density: 1.17 (20 °C)
Flash Pt: 122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007702
CAS Number: 137-07-5
EINECS: 205-277-3
UN: 3267
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/76/92/28927692.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR