2-Amino-9,10-anthraquinone ≥80.0% (by HPLC)

Synonyms: 2-Aminoanthraquinone

A0255-25G A0255-500G
TCA0255-500GEA 94.86 USD
TCA0255-500G TCA0255-025G
2-Amino-9,10-anthraquinone ≥80.0% (by HPLC)
2-Amino-9,10-anthraquinone

CAS Number: 117-79-3
MDL Number: MFCD00001233
Molecular Formula: C14H9NO2
Molecular Weight: 223.23
Purity/Analysis Method: 80.0% (HPLC)
Form: Crystal
Melting point (°C): 302
Flash Point (°C): 283

Formula: C₁₄H₉NO₂
MW: 223.23 g/mol
Boiling Pt: 473 °C (1013 hPa)
Melting Pt: 302 °C
Density: 1.45 g/cm³ (20 °C)
Flash Pt: 283 °C
MDL Number: MFCD00001233
CAS Number: 117-79-3
EINECS: 204-208-4
https://us.vwr.com/stibo/web/std.lang.all/76/81/28927681.jpg

Order Now

Specification Test Results

Purity/Analysis Method 80.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR