Adipamide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

A0160-25G
TCA0160-025GEA 86.56 USD
TCA0160-025G
Adipamide ≥98.0% (by total nitrogen basis)
Adipamide

CAS Number: 628-94-4
MDL Number: MFCD00008045
Molecular Formula: C6H12N2O2
Molecular Weight: 144.17
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 229


Other Product Names / Synonyms: Hexanediamide

Formula: C₆H₁₂N₂O₂
Melting Pt: 229 °C
MDL Number: MFCD00008045
CAS Number: 628-94-4
EINECS: 211-062-5
https://us.vwr.com/stibo/web/std.lang.all/76/02/28927602.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR