Acridine orange (hemizinc salt)

Danger

Synonyms: Basic Orange 143,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt)

A0132-25G
TCA0132-025GEA 55.62 USD
TCA0132-025G
Acridine orange (hemizinc salt)
Acridine orange (hemizinc salt)

CAS Number: 10127-02-3
MDL Number: MFCD00081043
Molecular Weight: 438.10
Form: Crystal


Other Product Names / Synonyms: Acridine Orange, Euchrysin GNY

Formula: C₃₄H₄₀Cl₄N₆Zn
MW: 369.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS Number: 10127-02-3
EINECS: 233-353-6
https://us.vwr.com/stibo/web/std.lang.all/75/80/28927580.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR