Acetylcholine iodide ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(2-Acetoxyethyl)trimethylammonium iodide
, 2-Acetoxy-N,N,N-trimethylethan-1-aminium iodide
A0085-25G
TCA0085-025GEA
107.03
USD
TCA0085-025G
Acetylcholine iodide ≥98.0% (by titrimetric analysis)
Acetylcholine iodide
CAS Number: 2260-50-6
MDL Number: MFCD00011815
Molecular Formula: C7H16INO2
Molecular Weight: 273.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 162
Formula:
H₃CC(O)O(CH₂)₂N(CH₃)₃I MW: 273.11 g/mol Melting Pt: 162 °C Density: 1.67 g/cm³ (20 °C) Storage Temperature: Freezer |
MDL Number:
MFCD00011815 CAS Number: 2260-50-6 EINECS: 218-862-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...