4-Nitrophenyl acetate ≥98.0%

Supplier: TCI America

Synonyms: 1-Acetoxy-4-nitrobenzene

A0040-100G A0040-25G
TCA0040-100GEA 558.25 USD
TCA0040-100G TCA0040-025G
4-Nitrophenyl acetate ≥98.0%
4-Nitrophenyl acetate

CAS Number: 830-03-5
MDL Number: MFCD00007326
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 78
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Acetic Acid 4-Nitrophenyl Ester

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Melting Pt: 78 °C
MDL Number: MFCD00007326
CAS Number: 830-03-5
EINECS: 212-593-5
https://us.vwr.com/stibo/web/std.lang.all/74/85/28927485.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR