Select™ Laboratory and Pharmacy Freezers, Nor-Lake® Scientific
NSSF241WWW/0 NSSF803WWW/0 NSSF522WWW/0 NSSF241WWW/4 NSSF331WWW/0 NSSF241WWW/5
89368-320EA 11998.34 USD
89368-320 89368-364 89368-322 89368-316 89368-318 89368-372
Select™ Laboratory and Pharmacy Freezers, Nor-Lake® Scientific
Laboratory and Pharmacy freezers are equipped with digital 4 x 20 character LCD display and microprocessor temperature controller. Freezers offer an adjustable temperature control range from –10 to –25 °C (14 to –13 °F). All units also provide 100-event alarm logging, integrated Hi/Lo product temperature alarm tests, digital calibration (air and product), real-time clock with date and time display, and a door opening counter with 24 hour automatic reset.
  • Alarms are adjustable, and may be set to beep or constant (keypad selectable)
  • Cabinet and solid door are foamed-in-place with CFC-free, high-density polyurethane foam insulation
  • The exterior cabinet front, sides, back, interior cabinet liner and interior door panel are painted white
  • Exterior cabinet top and bottom are galvanized steel
Standard safety features on all models include, remote alarm contacts, continuous product temperature display, three-sensor system (two product, one air), power supply switch, door ajar alarm (audible and visual), audible and visual Hi/Lo air and product temperature alarms, 2 to 10 volt DC output for product temperature (selectable to air temperature), and password protection set-points
Additional standard features include four casters on one and two door models (2 locking), six casters (3 locking) on three door models, three coated wire shelves per door section, interior light (switch activated), air cooled condensing unit, top-mounted forced air refrigeration system, automatic defrost, and condensate evaporation. The heavy-duty solid door(s) feature pivot hinges, full-length pull door handle(s), magnetic door gaskets, and centered key door lock(s) (2 keys included). Freezers include a 2.5 cm (1") diameter sensor access port.
Freezers are also equipped with standard monitoring features, including condenser/compressor run-time monitor, fan motor run-time monitor, and defrost heater time monitor.
Certifications: UL and C-UL listed.
Delivery information: Freezers are supplied with 24-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International). The 115V freezer 89368-316 is supplied with a 5-15P NEMA plug and 5-15R NEMA receptacle. The 115V freezers 89368-318, -320, and -322 are supplied with a 5-20P NEMA plug and 5-20R NEMA receptacle. The 208/230V freezers 89368-364, -366, -368, and -370 are supplied with a 6-15P NEMA plug and 6-15R NEMA receptacle. The 230V freezers 89368-372, -374, -376, and -378 feature a Power Inlet (IEC60320) module plug and require a receptacle cord supplied locally.