Would you like to log in again?
Premier™ Solid Door Low Temperature Laboratory Freezer, –40˚C, Nor-Lake® Scientific
Supplier: NOR-LAKE SCIENTIFIC
Premier™
NSXF221WWW/0
Freezer is equipped with a digital LED Microprocessor temperature controller with Hi/Lo audible and visual alarm, remote alarm contacts, and adjustable temperature control from –30 to –40˚C (–22 to –40°F)
Temperature is factory preset at –35˚C (–31°F). Standard safety features include, continuous product temperature display, product sensor with glycerin bottle, and power supply switch. The exterior cabinet front, sides, and back are painted white. Exterior cabinet top and bottom are galvanized steel. Interior cabinet liner and interior door panel are painted white. Cabinet and solid door are foamed-in-place with CFC-free, high-density polyurethane foam insulation. The heavy-duty door features pivot hinges, centered key door lock (2 keys included), and full-length door pull handle. Additional standard features include, four casters (2 locking), three epoxy-coated wire shelves, perimeter anti-condensate door frame (with temperature selectable heater wires), magnetic door gasket, air cooled condensing unit, and top-mounted forced air refrigeration system. Automatic defrost and condensate evaporation.
Certifications: UL and C-UL listed.
Ordering information: Freezers are supplied with an 18-month parts and labor warranty, 5-year compressor warranty (US and Canada), and 18-month parts warranty (International).
ORDER
Expand All / Collapse All
Description Electrical Volume Dimensions No. of Doors Temperature Range Weight Supplier No. VWR Catalog Number Unit Price Quantity
Premier™ Solid Door Low Temperature Laboratory Freezer 115V, 60Hz, 1PH 623 L (22 cu. ft.) 69.8W x 88.5D x 207H cm (271/2 x 347/8 x 811/2") 1 Solid Door –40 to –30°C (–40 to –22°F) 220 kg (485 lbs.) NSXF221WWW/0 89368-204 Each Retrieving
Product and/or packaging contains recycled/renewable content
Product and/or packaging is safer to human and environmental health UL Listed cUL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
SPECIFICATIONS
Environmentally Preferable Attribute Product and/or packaging contains recycled/renewable content, Product and/or packaging is safer to human and environmental health
SERVICES
Expand All / Collapse All
VWR® Customer On-site Calibration, Validation and PM Services
Description Details Includes Service Coverage Out-of-Service Coverage Supplier No. VWR Catalog Number Unit Price Quantity
VWR® NIST On-Site Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. Calibration at a single point using NIST traceable standards. Upon completion, a calibration form will be provided with “as found” and “as left” measurements. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-880 Each Retrieving
VWR® NIST On-Site PM and Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. PM and Calibration service includes cleaning of seals, fans, and gaskets. The chamber is inspected interiorly and exteriorly. After PM, technicians will calibrate the chamber as described in Calibration section. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-888 Each Retrieving
VWR® NIST Traceable Validate (IOQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. Validation (IQ/OQ) includes a standardized protocol procedure, calibration, tuning, IQ (verify design spec, system SOP, environmental conditions etc), and OQ (Perform empty chamber temperature mapping, open door and power loss testing). (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-950 Each Retrieving
VWR® NIST Traceable Validate (IOPQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. IQ/OQ will be performed as described in the (IQ/OQ) section. The PQ consists of loaded chamber temperature mapping for an additional 24 hours. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote.   75875-942 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty