4-Phenyl-1,2,3-thiadiazole

Warning

L20197-MD L20197-MD L20197-06 L20197-06 L20197-03 L20197-03
AAAL20197-03EA 127.84 USD
CAAAAL20197-06 AAAL20197-03 CAAAAL20197-03 AAAL20197-MD AAAL20197-06 CAAAAL20197-MD
4-Phenyl-1,2,3-thiadiazole
4-Phenyl-1,2,3-thiadiazole
Formula: C₈H₆N₂S
MW: 162.22 g/mol
Boiling Pt: ca 100 °C/1.5 mm
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00087232
CAS Number: 25445-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR