(S)-(-)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(-)-1-(4-Bromophenyl)ethylamine(S)-1-(4-Bromophenyl)ethanamine

L20080-06 L20080-03 L20080-14 L20080-06 L20080-14 L20080-03
AAAL20080-06EA 307.99 USD
CAAAAL20080-03 CAAAAL20080-14 AAAL20080-06 AAAL20080-14 AAAL20080-03 CAAAAL20080-06
(S)-(-)-4-Bromo-α-methylbenzylamine 99%, ee 98%, ChiPros®
(S)-(-)-4-Bromo-α-methylbenzylamine
Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 140…145 °C/30 mm
Melting Pt: –25 °C
Density: 1.371
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066026
CAS Number: 27298-97-1
EINECS: 608-076-2
UN: 2735
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR