(S)-(-)-Tetrahydro-2-furoic acid ≥99%, ee 98%

Danger

Synonyms: (2S)-(-)-Tetrahydrofuran-2-carboxylic acid(S)-Tetrahydrofuran-2-carboxylic acid

L19411-03 L19411-14 L19411-06 L19411-06 L19411-03
AAAL19411-06EA 133.78 USD
CAAAAL19411-06 AAAL19411-06 CAAAAL19411-03 AAAL19411-03 AAAL19411-14
(S)-(-)-Tetrahydro-2-furoic acid ≥99%, ee 98%
(S)-(-)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
MW: 116.12 g/mol
Boiling Pt: 244 °C
Density: 1.213
Flash Pt: 139 °C(282 °F)
Storage Temperature: Ambient
MDL Number: MFCD00272639
CAS Number: 87392-07-2
UN: 3265
ADR: "8",III

Order Now

Specification Test Results

Optical rotation -4° (c=1 in methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR