(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride 95%
L18336-MD
L18336-03
L18336-MD
L18336-03
AAAL18336-03EA
231.99
USD
CAAAAL18336-03
AAAL18336-03
CAAAAL18336-MD
AAAL18336-MD
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride 95%
(±)-3-Amino-4,4,4-trifluorobutyric acid hydrochloride
Formula:
C₄H₇ClF₃NO₂ Melting Pt: 197…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00181951 CAS Number: 91291-66-6 EINECS: 000-000-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...