3-Fluoro-L-tyrosine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-2-Amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid3-Fluoro-4-hydroxy-L-phenylalanine

L16775-MF L16775-MF
AAAL16775-MFEA 230.99 USD
AAAL16775-MF CAAAAL16775-MF
3-Fluoro-L-tyrosine 97%
3-Fluoro-L-tyrosine
Formula: C₉H₁₀FNO₃
MW: 199.18 g/mol
Melting Pt: 260…261 °C
Storage Temperature: Ambient
MDL Number: MFCD00002607
CAS Number: 7423-96-3
UN: 2811
ADR: "6.1",III

Order Now

Specification Test Results

Optical rotation -7° (c=1 in 1M HCl)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR