Triethylborane 1 M in THF
Supplier: Thermo Scientific Chemicals
|
Danger
|
L15109-AC
L15109-AE
L15109-AE
L15109-AC
AAAL15109-AEEA
160.99
USD
CAAAAL15109-AC
AAAL15109-AE
CAAAAL15109-AE
AAAL15109-AC
Triethylborane 1 M in THF
Triethylborane
Formula:
C₆H₁₅B Density: 0.865 Flash Pt: -17 °C(1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009022 CAS Number: 97-94-9 EINECS: 202-620-9 UN: 2924 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...