(R)-(+)-4-Isopropyl-2-oxazolidinone 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(4R)-(+)-4-Isopropyl-2-oxazolidinone
, (R)-4-Isopropyloxazolidin-2-one
L14518-03
L14518-03
L14518-MD
AAAL14518-MDEA
33.85
USD
AAAL14518-MD
AAAL14518-03
CAAAAL14518-03
(R)-(+)-4-Isopropyl-2-oxazolidinone 98+%
(R)-(+)-4-Isopropyl-2-oxazolidinone
Formula:
C₆H₁₁NO₂ MW: 129.16 g/mol Melting Pt: 70…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075172 CAS Number: 95530-58-8 |
Specification Test Results
Optical rotation | +17° (c=6 in ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...