Glycine octyl ester hydrochloride 98%
Synonyms:
Octyl 2-aminoacetate hydrochloride
, Glycine n-octyl ester hydrochloride
L13385-06
L13385-03
L13385-03
L13385-06
AAAL13385-03EA
44.99
USD
CAAAAL13385-06
AAAL13385-03
CAAAAL13385-03
AAAL13385-06
Glycine octyl ester hydrochloride 98%
Glycine octyl ester hydrochloride
Formula:
C₁₀H₂₂ClNO₂ MW: 223.75 g/mol Melting Pt: 66…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151827 CAS Number: 39540-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...