1-Bromo-3,3,4,4,4-pentafluoro-2-butanone 97%

Danger

L12199-06 L12199-06
AAAL12199-06EA 290.84 USD
AAAL12199-06 CAAAAL12199-06
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone 97%
1-Bromo-3,3,4,4,4-pentafluoro-2-butanone
Formula: C₄H₂BrF₅O
MW: 240.96
Boiling Pt: 93…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00041447
CAS Number: 92737-01-4
EINECS: 000-000-0
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR