Triethyl-1,1,2-ethanetricarboxylate 99%

Synonyms: Triethyl ethane-1,1,2-tricarboxylate

L12014-14 L12014-14 L12014-06
AAAL12014-14EA 82.11 USD
CAAAAL12014-06 AAAL12014-14 CAAAAL12014-14
Triethyl-1,1,2-ethanetricarboxylate 99%
Triethyl 1,1,2-ethanetricarboxylate
Formula: C₁₁H₁₈O₆
MW: 246.26 g/mol
Boiling Pt: 104…105 °C/1 mm
Density: 1,074
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009154
CAS Number: 7459-46-3
EINECS: 231-235-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR