(R)-(-)-Prolinol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
D-Prolinol
, D(-)-Prolinol
, (2R)-pyrrolidin-2-ylmethanol
L11109-14
L11109-03
L11109-06
L11109-03
AAL11109-06EA
278.99
USD
CAAAAL11109-03
AAL11109-06
AAAL11109-03
AAL11109-14
(R)-(-)-Prolinol 98+%
(R)-(-)-Prolinol
Formula:
C₅H₁₁NO MW: 101.1 g/mol Boiling Pt: 74…76 °C/2 mm Density: 1.025 Flash Pt: 86 °C(186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064321 CAS Number: 68832-13-3 EINECS: 629-147-4 |
Specification Test Results
Optical rotation |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...