Methyl-3-chlorobenzo[b]thiophene-2-carboxylate 95%
|
Warning
|
Synonyms:
Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
L10696-06
L10696-03
L10696-06
L10696-03
AAAL10696-06EA
167.46
USD
CAAAAL10696-06
AAAL10696-06
AAAL10696-03
CAAAAL10696-03
Methyl-3-chlorobenzo[b]thiophene-2-carboxylate 95%
Methyl 3-chlorobenzo[b]thiophene-2-carboxylate
Formula:
C₁₀H₇ClO₂S MW: 226.68 g/mol Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052503 CAS Number: 21211-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...