Methyl butyrylacetate 98+%
Warning
|
Synonyms:
Methyl 3-oxohexanoate
L09826-22
L09826-22
L09826-14
L09826-14
AAAL09826-22EA
74.8
USD
AAAL09826-22
CAAAAL09826-22
CAAAAL09826-14
AAAL09826-14
Methyl butyrylacetate 98+%
Methyl butyrylacetate
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol Boiling Pt: 85…90 °C/5 mm Melting Pt: –43 °C Density: 1,037 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040995 CAS Number: 30414-54-1 EINECS: 250-185-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...