Tetraethylurea 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,3,3-Tetraethylurea

L09458-14 L09458-06
AAAL09458-14EA 100 USD
AAAL09458-14 AAAL09458-06
Tetraethylurea 99%
Tetraethylurea
Formula: C₉H₂₀N₂O
MW: 172.27
Boiling Pt: 212…214 °C
Density: 0,910
Flash Pt: 81 °C(177 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042881
CAS Number: 1187-03-7
EINECS: 214-696-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR