2-Thienylacetyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Thiophene-2-acetyl chloride
, 2-Thiopheneacetyl chloride
L09323-14
L09323-14
AAAL09323-14EA
79.59
USD
CAAAAL09323-14
AAAL09323-14
2-Thienylacetyl chloride 98%
2-Thienylacetyl chloride
Formula:
C₆H₅ClOS MW: 160.62 Boiling Pt: 131…132 °C/90 mm Density: 1.303 Flash Pt: 101 °C(213 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005456 CAS Number: 39098-97-0 EINECS: 254-290-0 UN: 1760 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...