4-Nonyloxybenzaldehyde 98%
L09110-04
L09110-04
L09110-09
L09110-09
AAAL09110-04EA
43.06
USD
AAAL09110-04
CAAAAL09110-04
CAAAAL09110-09
AAAL09110-09
4-Nonyloxybenzaldehyde 98%
4-Nonyloxybenzaldehyde
Formula:
C₁₆H₂₄O₂ Boiling Pt: 180…182 °C/4 mm Storage Temperature: Ambient |
MDL Number:
MFCD00043712 CAS Number: 50262-46-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...