7-Benzoylenanthic acid 98+%
|
Warning
|
Synonyms:
7-Benzoylheptanoic acid
L08395-03
L08395-06
L08395-06
L08395-03
AAAL08395-06EA
176.49
USD
CAAAAL08395-06
AAAL08395-06
CAAAAL08395-03
AAAL08395-03
7-Benzoylenanthic acid 98+%
7-Benzoylenanthic acid
Formula:
C₁₄H₁₈O₃ Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039609 CAS Number: 66147-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...