3-Chloro-L-tyrosine 97%

Synonyms: H-Tyr(3-Cl)-OH

L07675-03 L07675-06 L07675-MD L07675-MD L07675-06
AAAL07675-MDEA 32.88 USD
CAAAAL07675-03 AAAL07675-MD AAAL07675-06 CAAAAL07675-06 CAAAAL07675-MD
3-Chloro-L-tyrosine 97%
3-Chloro-L-tyrosine
Formula: C₉H₁₀ClNO₃
MW: 215.7 g/mol
Melting Pt: ca 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00045865
CAS Number: 7423-93-0
EINECS: 231-050-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR