2-(Ethylthio)ethanol 96%

Danger

Synonyms: 2-(Ethylmercapto)-ethanolEthyl 2-hydroxyethyl sulfideEthyl 2-hydroxyethyl sulphide

L05040-18 L05040-18
AAAL05040-18EA 352.69 USD
CAAAAL05040-18 AAAL05040-18
2-(Ethylthio)ethanol 96%
2-(Ethylthio)ethanol
Formula: C₄H₁₀OS
Boiling Pt: 180…184 °C
Melting Pt: ca …100 °C
Density: 1.018
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002909
CAS Number: 110-77-0
EINECS: 203-802-0
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR