3-(4-Bromobenzoyl)propionic acid 98+%
|
Warning
|
Synonyms:
4-(4-Bromophenyl)-4-oxobutanoic acid
L04008-03
L04008-06
L04008-14
L04008-06
L04008-14
L04008-03
AAAL04008-14EA
223.11
USD
AAAL04008-14
AAAL04008-03
CAAAAL04008-03
CAAAAL04008-14
AAAL04008-06
CAAAAL04008-06
3-(4-Bromobenzoyl)propionic acid 98+%
3-(4-Bromobenzoyl)propionic acid
Formula:
C₁₀H₉BrO₃ MW: 257.08 g/mol Melting Pt: 145…148 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016563 CAS Number: 6340-79-0 EINECS: 228-732-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...