4-Fluorophenylurea 96%
Supplier: Thermo Scientific Chemicals
L01314-06
L01314-14
L01314-14
L01314-06
AAAL01314-06EA
46.69
USD
AAAL01314-06
CAAAAL01314-06
AAAL01314-14
CAAAAL01314-14
4-Fluorophenylurea 96%
4-Fluorophenylurea
Formula:
C₇H₇FN₂O Melting Pt: 183…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014787 CAS Number: 659-30-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...