N-(3-Pyridyl)thiourea 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(3-Pyridyl)-2-thiourea
L01284-06
L01284-06
L01284-03
L01284-03
AAAL01284-03EA
77.79
USD
CAAAAL01284-03
AAAL01284-03
CAAAAL01284-06
AAAL01284-06
N-(3-Pyridyl)thiourea 98+%
N-(3-Pyridyl)thiourea
Formula:
C₆H₇N₃S Melting Pt: 158…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014635 CAS Number: 30162-37-9 EINECS: 250-075-0 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...