3-Methyl-5-phenyl-1H-pyrazole
Supplier: Thermo Scientific Chemicals
H50650-03
H50650-03
H50650-06
H50650-06
AAAH50650-03EA
138.99
USD
AAAH50650-03
CAAAAH50650-06
AAAH50650-06
CAAAAH50650-03
3-Methyl-5-phenyl-1H-pyrazole
3-Methyl-5-phenyl-1H-pyrazole
Formula:
C₁₀H₁₀N₂ MW: 158.2 g/mol Melting Pt: 124…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00524752 CAS Number: 3347-62-4 EINECS: 222-099-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...