4-(Boc-amino)-2-chloropyridine 95%
|
Warning
|
Synonyms:
tert-Butyl (2-chloropyridin-4-yl)carbamate
, 4-Amino-2-chloropyridine, 4-BOC protected
, 2-Chloro-4-(tert-butoxycarbonyl)pyridine
, tert-Butyl 2-chloropyridine-4-carbamate
H50067-06
H50067-06
H50067-03
H50067-03
AAAH50067-06EA
863.12
USD
AAAH50067-06
CAAAAH50067-06
AAAH50067-03
CAAAAH50067-03
4-(Boc-amino)-2-chloropyridine 95%
4-(Boc-amino)-2-chloropyridine
Formula:
C₁₀H₁₃ClN₂O₂ MW: 228.68 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02179232 CAS Number: 234108-73-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...