Perfluorobutane-1-sulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nonafluorobutane-1-sulphonyl chloride
H27437-03
H27437-06
AAH27437-06EA
472.95
USD
AAH27437-06
AAH27437-03
Perfluorobutane-1-sulfonyl chloride 98%
Perfluorobutane-1-sulphonyl chloride
Formula:
C₄ClF₉O₂S MW: 318.55 Storage Temperature: Ambient |
MDL Number:
MFCD03412262 CAS Number: 2991-84-6 UN: 3265 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...