2-Phenyl-9,10-anthraquinone 98%
Synonyms:
2-Phenylanthraquinone
H27376-MD
AAH27376-MDEA
38.46
USD
AAH27376-MD
2-Phenyl-9,10-anthraquinone 98%
2-Phenyl-9,10-anthraquinone
Formula:
C₂₀H₁₂O₂ Melting Pt: 160…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00559262 CAS Number: 6485-97-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...