D-α-Aminoadipic acid 98%
Synonyms:
(R)-2-Aminohexanedioic acid
, D-2-Aminoadipic acid
H27345-MD
AAH27345-MDEA
102.34
USD
AAH27345-MD
D-α-Aminoadipic acid 98%
D-α-Aminoadipic acid
Formula:
C₆H₁₁NO₄ MW: 161.16 g/mol Melting Pt: 205 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063118 CAS Number: 7620-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...