Ammonia 2 M in methanol

Supplier: Thermo Scientific Chemicals
Danger

H27080-AE H27080-AU
AAH27080-AEEA 107.45 USD
AAH27080-AE AAH27080-AU
Ammonia 2 M in methanol
Ammonia
Formula: NH₃
MW: 17.03 g/mol
Density: 0.787
Flash Pt: 6 °C(43 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011418
CAS Number: 7664-41-7
UN: 1992
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR