(S)-(-)-Indoline-2-carboxylic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-Indoline-2-carboxylic acid

H27009-03 H27009-06
AAH27009-06EA 129.99 USD
AAH27009-06 AAH27009-03
(S)-(-)-Indoline-2-carboxylic acid 97+%
(S)-(-)-Indoline-2-carboxylic acid
Formula: C₉H₉NO₂
MW: 163.18 g/mol
Melting Pt: ca 177 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00070578
CAS Number: 79815-20-6
EINECS: 410-860-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR