2-Chloro-3,6-difluoroaniline 97%

Danger

H26822-06 H26822-03
AAH26822-03EA 125.43 USD
AAH26822-03 AAH26822-06
2-Chloro-3,6-difluoroaniline 97%
2-Chloro-3,6-difluoroaniline
Formula: C₆H₄ClF₂N
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD06660176
CAS Number: 287172-80-9
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR