2-Chloro-5-fluorobenzonitrile 97%

Danger

H26817-06 H26817-03 H26817-14
AAH26817-03EA 59.26 USD
AAH26817-03 AAH26817-14 AAH26817-06
2-Chloro-5-fluorobenzonitrile 97%
2-Chloro-5-fluorobenzonitrile
Formula: C₇H₃ClFN
MW: 155.56 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD03094168
CAS Number: 57381-56-3
EINECS: 260-714-5
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR