3-Chloro-2-fluorophenol 97%
|
Danger
|
H26725-03
H26725-06
AAH26725-06EA
360.75
USD
AAH26725-06
AAH26725-03
3-Chloro-2-fluorophenol 97%
3-Chloro-2-fluorophenol
Formula:
C₆H₄ClFO MW: 146.55 g/mol Boiling Pt: 190 °C Melting Pt: 34…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631572 CAS Number: 2613-22-1 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...