(±)-4-Fluorostyrene oxide 98%

Warning

H26600-06 H26600-14
AAH26600-14EA 276.77 USD
AAH26600-14 AAH26600-06
(±)-4-Fluorostyrene oxide 98%
(±)-4-Fluorostyrene oxide
Formula: C₈H₇FO
Boiling Pt: 92 °C/14 mm
Density: 1.167
Flash Pt: 77 °C(171 °F)
Storage Temperature: Ambient
MDL Number: MFCD00043994
CAS Number: 18511-62-1
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR